missing translation for 'onlineSavingsMsg'
Learn More

Nitrite de sodium, 99,999 % (pureté du métal), Thermo Scientific Chemicals

Catalog Number p-4407921
Click to view available options
Quantité:
25 g
100 g
7632-00-0
NNaO2
69.00
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
23668193
CHEBI:78870
nitrite de sodium
[Na+].[O-]N=O
7632-00-0
NNaO2
69.00
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
23668193
CHEBI:78870
nitrite de sodium
[Na+].[O-]N=O

Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor and in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Sodium nitrite
Point de fusion 271°C
Couleur Blanc
Forme physique Crystals
Quantité 25 g
Plage de pourcentage du dosage 99.999% (metals basis)
Informations sur la solubilité Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether.
Poids de la formule 69
Odeur Odorless
Sensibilité Hygroscopic
Densité 2.17 g/cm3 at 20°C
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'