missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORLA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22592-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer′s Disease.Spécification
SORLA | |
Polyclonal | |
Unconjugated | |
SORL1 | |
LR11, LRP9, SORL1, SORLA, SorLA 1, Sortilin related receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20660, 300652, 6653 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88307, Q92673 | |
SORL1 | |
SORLA Fusion Protein Ag18445 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |