missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25415-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.Spécification
SOX15 | |
Polyclonal | |
Unconjugated | |
SOX15 | |
Protein SOX 12, Protein SOX 15, Protein SOX 20, SOX12, SOX15, SOX20, SOX26, SOX27 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20670, 363632, 6665 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60248, P43267 | |
Sox15 | |
SOX15 Fusion Protein Ag22032 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |