missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13216-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Spécification
SOX5 | |
Polyclonal | |
Unconjugated | |
SOX5 | |
L SOX5, SOX5, Transcription factor SOX 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140587, 20678, 6660 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35710, P35711 | |
SOX5 | |
SOX5 Fusion Protein Ag4092 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |