Learn More
SOX9 / SRY-box 9 Mouse, Clone: PCRP-SOX9-1E5, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 6662-MSM1-P1
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15980888
  • 485.00€ / 100µg
Expédition estimée: 24-01-2024
Spécification
SOX9 / SRY-box 9
Nucleus
PCRP-SOX9-1E5
Unconjugated
Mouse
65kDa
Protein A/G purified from Bioreactor concentrate.
RUO
6662
IgG2b
ELISA
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human SOX9 protein
Pancreas
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.