missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26584-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13.Spécification
SP3 | |
Polyclonal | |
Unconjugated | |
SP3 | |
SP3, Sp3 transcription factor, SPR 2, Transcription factor Sp3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20687, 6670 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70494, Q02447 | |
SP3 | |
SP3 Fusion Protein Ag25082 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |