missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | SPAST |
---|---|
Concentration | 0.44 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16842095
|
Proteintech
22792-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16832095
|
Proteintech
22792-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. The encoded ATPase may be involved in the assembly or function of nuclear protein complexes. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequences have not been determined. Mutations associated with this gene cause the most frequent form of autosomal domit spastic paraplegia 4.Spécification
SPAST | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
B2RYN7, Q9QYY8, Q9UBP0 | |
362700, 50850, 6683 | |
SPAST Fusion Protein Ag18797 | |
Primary | |
-20°C | |
SPAST |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ADPSP, FSP2, KIAA1083, SPAST, Spastic paraplegia 4 protein, spastin, SPG4 | |
Spast | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |