Learn More
Cole-Parmer™ CHRONO REBOURS 9999 MIN ALARME
Marque: Cole-Parmer™ 94460-42
Code nomenclature Nacres: NB.51
Informations supplémentaires : Poids : 0.80000kg
Spécification
Digital Timer | |
Audible/Visible | |
1 sec. | |
76.2 mm | |
50.8 mm | |
1 Each |
9999 min. | |
±3sec./day | |
Digital LCD | |
50.8 mm | |
Two AAA Batteries, Built-in Stand, Pocket Clip, and Magnetic Mounting Strips |