missing translation for 'onlineSavingsMsg'
Learn More

SRGAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22519-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801965

  • 329.85€ / 150µL

Expédition estimée
04-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
Spécification

Spécification

SRGAP2
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75044, Q91Z67
ARHGAP34, FNBP2, Formin binding protein 2, KIAA0456, SRGAP2, srGAP3
Rabbit
IgG
150 μL
-20°C
Primary
14270, 23380
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SRGAP2
Liquid
SRGAP2
SRGAP2 Fusion Protein Ag18278
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents