missing translation for 'onlineSavingsMsg'
Learn More

ST6GAL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14355-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802964

  • 329.85€ / 150µL

Expédition estimée
04-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Western blot analysis of human liver using this antibody detects both isoforms between 43-50 kDa. Higher molecular weight of bands around 50-70 kDa can also be observed with unknown reason.

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Spécification

Spécification

ST6GAL1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13721, P15907, Q64685
Alpha 2,6 ST 1, B cell antigen CD75, CD75, Sialyltransferase 1, SIAT1, ST6Gal I, ST6GAL1, ST6GalI, ST6N
Rabbit
IgG
150 μL
-20°C
Primary
20440, 25197, 6480
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ST6GAL1
Liquid
ST6GAL1
ST6GAL1 Fusion Protein Ag5705
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents