Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags, Coated wire holder for 509 & 510 bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Marque:  Thermo Scientific™ S23C
Code nomenclature Nacres: NB.87
Informations supplémentaires : Poids : 1.60000kg
Code produit. 11399103
Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
Voir les produits de remplacement »
Description
Description
Thermo Scientific™ Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Spécification
Spécification
Cardboard holder for 511 bags
Cardboard
1