missing translation for 'onlineSavingsMsg'
Learn More

STXBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15312-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16804444

  • 329.85€ / 150µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Spécification

Spécification

STXBP2
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15833, Q64324
Hunc18b, MUNC18 2, pp10122, STXBP2, STXBP2 protein, syntaxin binding protein 2, UNC18 2, UNC18B
Rabbit
IgG
150 μL
-20°C
Primary
20911, 6813
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STXBP2
Liquid
STXBP2
STXBP2 Fusion Protein Ag7537
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents