missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUCLG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14240-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a GTP-specific beta subunit of succinyl-CoA synthetase. Succinyl-CoA synthetase catalyzes the reversible reaction involving the formation of succinyl-CoA and succinate. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 5 and 12.Spécification
SUCLG2 | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96I99, Q9Z2I8 | |
G BETA, SCS betaG, SUCLG2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
20917, 362404, 8801 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
SUCLG2 | |
Liquid | |
Suclg2 | |
SUCLG2 Fusion Protein Ag5495 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |