missing translation for 'onlineSavingsMsg'
Learn More

SUPT6H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23073-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802185

  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody specifically recognizes the 230 kDa phosphorylated SUPT6H protein.

Transcription elongation factor which binds histone H3 and plays a key role in the regulation of transcription elongation and mRNA processing. Enhances the transcription elongation by RNA polymerase II (RNAPII) and is also required for the efficient activation of transcriptional elongation by the HIV-1 nuclear transcriptional activator, Tat. Besides chaperoning histones in transcription, acts to transport and splice mRNA by forming a complex with IWS1 and the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2), to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription. SUPT6H via its association with SETD1A, regulates both class-switch recombination and somatic hypermutation through formation of H3K4me3 epigenetic marks on activation-induced cytidine deaminase (AICDA) target loci. Promotes the activation of the myogenic gene program by entailing erasure of the repressive H3K27me3 epigenetic mark through stabilization of the chromatin interaction of the H3K27 demethylase KDM6A.
Spécification

Spécification

SUPT6H
Polyclonal
Unconjugated
SUPT6H
emb 5, hSPT6, KIAA0162, SPT6, SPT6H, SUPT6H, Tat cotransactivator 2 protein, Tat CT2 protein
Rabbit
Antigen Affinity Chromatography
RUO
20926, 303281, 6830
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62383, Q7KZ85
SUPT6H
SUPT6H Fusion Protein Ag19426
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions