missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin-11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12031-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.Spécification
Synaptotagmin-11 | |
Polyclonal | |
Unconjugated | |
SYT11 | |
KIAA0080, Synaptotagmin 11, synaptotagmin XI, Synaptotagmin-11, SYT11, SYT12, SytXI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229521, 23208, 60568 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08835, Q9BT88, Q9R0N3 | |
Syt11 | |
Synaptotagmin-11 Fusion Protein Ag2663 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |