missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaphilin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13646-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Syntaxin-1, synaptobrevin/VAMP, and SNAP25 interact to form the SNARE complex, which is required for synaptic vesicle docking and fusion. The protein encoded by this gene is membrane-associated and inhibits SNARE complex formation by binding free syntaxin-1. Expression of this gene appears to be brain-specific.Spécification
Syntaphilin | |
Polyclonal | |
Unconjugated | |
SNPH | |
KIAA0374, SNPH, syntaphilin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
241727, 296267, 9751 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B5DF41, O15079, Q80U23 | |
SNPH | |
Syntaphilin Fusion Protein Ag4594 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |