missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13301-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the syntaxin family. Syntaxins have been implicated in the targeting and fusion of intracellular transport vesicles. This family member may regulate protein transport among late endosomes and the trans-Golgi network. Mutations in this gene have been associated with familial hemophagocytic lymphohistiocytosis.Spécification
Syntaxin 11 | |
Polyclonal | |
Unconjugated | |
STX11 | |
FHL4, HLH4, HPLH4, STX11, syntaxin 11, Syntaxin 11,SYX11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292483, 74732, 8676 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75558, Q9D3G5 | |
Stx11 | |
Syntaxin 11 Fusion Protein Ag4097 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |