missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syrian Hamster IgG Isotype Control, Invitrogen™
Hamster Isotype Control
Marque: Invitrogen 31246
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity/Antibody specificity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against rabbit anti-Syrian hamster whole serum is the same as that against rabbit anti-Syrian hamster IgG, Fc fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Store at 4oC under sterile conditions. Dilute only enough product for a single day’s use. Country of Origin: USA
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Spécification
Syrian Hamster IgG | |
5.9 mg/mL | |
PBS with no preservative; pH 7.6 | |
Hamster | |
5 mg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry | |
Unconjugated | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |