Learn More
TCF12/HEB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14419-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.Spécification
TCF12/HEB | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51514, Q61286, Q99081 | |
bHLHb20, DNA binding protein HTF4, E box binding protein, HEB, HsT17266, HTF4, TCF 12, TCF12, transcription factor 12, Transcription factor HTF 4 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
21406, 25720, 6938 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TCF12 | |
Liquid | |
TCF12 | |
TCF12/HEB Fusion Protein Ag5765 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Rat, Mouse, Human |