missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Spécification
Antigène | TEKT1 |
---|---|
Concentration | 0.38 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16812424
|
Proteintech
13969-1-AP-20UL |
20 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
16802424
|
Proteintech
13969-1-AP-150UL |
150 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predomitly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.Spécification
TEKT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q969V4, Q99JD2, Q9DAJ2 | |
21689, 83659, 85270 | |
TEKT1 Fusion Protein Ag4993 | |
Primary | |
-20°C | |
TEKT1 |
0.38 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TEKT1 | |
Tekt1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |