missing translation for 'onlineSavingsMsg'
Learn More

TEKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13969-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802424

  • 329.85€ / 150µL

Expédition estimée
08-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predomitly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.
Spécification

Spécification

TEKT1
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969V4, Q99JD2, Q9DAJ2
TEKT1
Rabbit
IgG
150 μL
-20°C
Primary
21689, 83659, 85270
Western Blot
Unconjugated
TEKT1
Liquid
Tekt1
TEKT1 Fusion Protein Ag4993
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents