Learn More
TELO2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15975-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16805404
  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
Description
TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.
Spécification
Spécification
TELO2
Polyclonal
Unconjugated
TELO2
CLK2, hCLK2, KIAA0683, Protein clk 2 homolog, TEL2, TELO2
Rabbit
Antigen Affinity Chromatography
RUO
9894
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y4R8
TELO2
TELO2 Fusion Protein Ag8763
150 μL