missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solution d’acide tétrafluoroborique, Honeywell Fluka™
Click to view available options
Quantité:
25 g
500 g
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Nom chimique ou matériau | Tetrafluoroboric Acid Solution |
Concentration | 48wt. % in H2O |
Densité | 1.4g/mL at 25°C |
Tension de vapeur | 5mmHg (20°C) |
Quantité | 25 g |
Formule linéaire | HBF4 |
Numéro UN | 1775 |
Poids de la formule | 87.81g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'