missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB2M Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24411-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
S-adenosyl-L-methionine-dependent methyltransferase which specifically dimethylates mitochondrial 12S rRNA at the conserved stem loop. Also required for basal transcription of mitochondrial DNA, probably via its interaction with POLRMT and TFAM. Stimulates transcription independently of the methyltransferase activity. Compared to TFB1M, it activates transcription of mitochondrial DNA more efficiently, while it has less methyltransferase activity.Spécification
TFB2M | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3TL26, Q9H5Q4 | |
h mtTFB, h mtTFB2, Hkp1, hTFB2M, mtTFB2, NS5ATP5, TFB2M | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
15278, 64216 |
Western Blot | |
Unconjugated | |
TFB2M | |
Liquid | |
TFB2M | |
TFB2M Fusion Protein Ag19687 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |