missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ PepMap™100 Fusica™ Capillary Columns, 5μm Particle Size

Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples

Marque:  Thermo Scientific™ 164899

Code nomenclature Nacres: SB.52

Informations supplémentaires : Poids : 0.50000kg

 Afficher plus de versions de ce produit

Code produit. 15122428

  • 825.00€ / 1 pièce


Date dexpédition estimée
du fournisseur 26-07-2018
Ajouter au panier

Description

Description

  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC

  • Inside diameter 300μm

Spécification

Spécification

1000μm
50mm
5μm
Acclaim PepMap C18 100A nanoViper HPLC Column
Vidéos