Learn More
Thermo Scientific™ Buffers for MagJET™ Blood gDNA Kit
Marque:  Thermo Scientific™ R2342
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 0.04000kg
Code produit. 13565190
  • 137.00€ / 60mL
Date dexpédition estimée
du fournisseur 30-07-2018
Ajouter au panier
Description
Description
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Spécification
Spécification
MagJET Blood gDNA Kit
60mL
Vidéos