Learn More
Thermo Scientific™ Buffers for MagJET™ Plant gDNA Kit
Marque:  Thermo Scientific™ R2492
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 0.07000kg
Code produit. 13575210
  • 123.00€ / 32mL
Date dexpédition estimée
du fournisseur 30-07-2018
Ajouter au panier
Description
Description
The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Spécification
Spécification
MagJET Plant gDNA Kit
32mL
Vidéos