Learn More
Thermo Scientific™ Buffers for MagJET™ Viral Kit
Marque:  Thermo Scientific™ R2661
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 0.12000kg
 Afficher plus de versions de ce produit
Code produit. 13525240
  • 41.95€ / 50mL
Date dexpédition estimée
du fournisseur 08-04-2019
Ajouter au panier
Description
Description
The Thermo Scientific™ MagJET™ Viral DNA and RNA Kit is designed for fast and efficient purification of viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA and RNA is free of proteins, nucleases and other contaminants or inhibitors.
Spécification
Spécification
MagJET Viral Kit
50mL
Vidéos