Learn More
Thermo Scientific™ MagJET™ RNA Kit
Uses magnetic bead technology to purify total RNA from cultured mammalian cells, tissues, bacteria, and yeast.
Marque:  Thermo Scientific™ K2731
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 2.31330kg
Code produit. 13555170
  • 356.00€ / 96 tests
Date dexpédition estimée
du fournisseur 02-04-2019
Ajouter au panier
Description
Description
Kits de purification d'acides nucléiques MagJET
• Basé sur le système de billes magnétiques
• Idéal pour automates
• Grande pureté des acides nucléiques permettant l'utilisation direct dans les applications ultérieures
• Kit MagJET pour ARN
Spécification
Spécification
High purity RNA suitable for all conventional molecular biology procedures like RT-PCR, RT-qPCR, Northern blotting, Nuclease protection assay
Lysis buffer (50mL), MagJET magnetic beads (3 x 1.4mL), lyophilized DNase I (1 vial), 10x reaction buffer with MgCl2 for DNase I (3 x 1mL), Conc. DNase I reconstitution buffer (1mL), conc. wash buffer 1 (40mL), conc. wash buffer 2 (45mL), nuclease free water (30mL)
96 Preparations
Store lyophilized DNase I, 10x Reaction Buffer with MgCl2 for DNase I and DNase I Reconstitution Buffer at -20deg.C upon arrival. Store MagJET Magnetic Beads at 4deg.C and other components of the kit at room temperature (15deg. to 25deg.C)
Vidéos
Documentation
Documents techniques