Learn More
Thermo Scientific™ Reagents for EpiJET™ Bisulfite Conversion Kit
Marque:  Thermo Scientific™ R2151
Code nomenclature Nacres: NA.53
Informations supplémentaires : Poids : 0.05000kg
Code produit. 13515160
  • 49.33€ / 3.50mL
Date dexpédition estimée
du fournisseur 06-06-2018
Ajouter au panier
Description
Description
The EpiJET Bisulfite Conversion Kit is designed for simple and reliable bisulfite conversion of DNA for methylation analysis. The kit integrates the thermal DNA denaturation and the bisulfite conversion reactions into one step. The unmethylated cytosines are deaminated and converted to uracils using powdered Modification Reagent that has been reconstituted with the two Modification Solutions. This step can be performed following long or short protocol, depending on the downstream application. The converted DNA is bound to a membrane for on-column desulfonation and subsequent purification steps, and then eluted. The resulting elution of the converted DNA is suitable for a number of techniques used in methylation status analysis, including PCR, qPCR, COBRA and sequencing.
Spécification
Spécification
3.5mL
EpiJET Bisulfite Conversion Kit
Vidéos