Learn More
Thermo Scientific™ Reagents for MagJET™ gDNA Kit
Marque:  Thermo Scientific™ R2461
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 0.07000kg
Code produit. 13525210
  • 48.51€ / 25mL
Date dexpédition estimée
du fournisseur 08-04-2019
Ajouter au panier
Description
Description
The Thermo Scientific™ MagJET™ Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Spécification
Spécification
MagJET gDNA Kit
25mL
Vidéos