Learn More
Thermo Scientific™ Reagents for MagJET™ Plasmid DNA Kit
Marque:  Thermo Scientific™ R2632
Code nomenclature Nacres: NA.52
Informations supplémentaires : Poids : 0.05000kg
Code produit. 13575230
  • 66.75€ / 45mL
Date dexpédition estimée
du fournisseur 08-04-2019
Ajouter au panier
Description
Description
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Spécification
Spécification
MagJET Plasmid DNA Kit
45mL
Vidéos