missing translation for 'onlineSavingsMsg'
Learn More

TIM-3 (CD366) Mouse anti-Human, Alexa Fluor 488, Clone: 7D3, BD Biosciences™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 567031

Informations supplémentaires : Poids : 0.00020kg

 Afficher plus de versions de ce produit

Code produit. 17257814

  • 202.00€ / 100µg
Expédition estimée: 22-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry
Spécification

Spécification

TIM-3 (CD366)
Monoclonal
Alexa Fluor 488
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
Human TIM-3
RUO
Human
IgG1 κ
Flow Cytometry
7D3
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions