missing translation for 'onlineSavingsMsg'
Learn More

TLR6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22240-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801785

  • 329.85€ / 150µL

Expédition estimée
04-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This receptor functionally interacts with toll-like receptor 2 to mediate cellular response to bacterial lipoproteins.
Spécification

Spécification

TLR6
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPW9, Q9Y2C9
CD286, TLR6, toll like receptor 6
Rabbit
IgG
150 μL
-20°C
Primary
10333, 21899
Western Blot
Unconjugated
TLR6
Liquid
TLR6
TLR6 Fusion Protein Ag17686
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents