missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM127 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23142-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
TMEM127 controls cell proliferation acting as a negative regulator of TOR signaling pathway mediated by mTORC1. May act as a tumor suppressor.Spécification
TMEM127 | |
Polyclonal | |
Unconjugated | |
TMEM127 | |
TMEM127, transmembrane protein 127 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55654, 69470 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75204, Q8BGP5 | |
TMEM127 | |
TMEM127 Fusion Protein Ag19472 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |