missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM132A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25301-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that is highly similar to the rat Grp78-binding protein (GBP). Alternatively spliced transcript variants encoding different isoforms have been described.Spécification
TMEM132A | |
Polyclonal | |
Unconjugated | |
TMEM132A | |
GBP, HSPA5 binding protein 1, HSPA5BP1, KIAA1583, TMEM132A, transmembrane protein 132A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54972, 98170 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q24JP5, Q922P8 | |
TMEM132A | |
TMEM132A Fusion Protein Ag18142 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |