missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM139 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21912-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
TMEM139 | |
Polyclonal | |
Unconjugated | |
TMEM139 | |
TMEM139, transmembrane protein 139 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109218, 135932 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IV31 | |
Tmem139 | |
TMEM139 Fusion Protein Ag15354 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |