Learn More
TMEM141 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16092-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16805584
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
Description
TMEM141 is a protein coding gene.
Spécification
Spécification
TMEM141
Polyclonal
Unconjugated
TMEM141
RP11 216L13.7, TMEM141, transmembrane protein 141
Rabbit
Antigen Affinity Chromatography
RUO
499755, 51875, 85014
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2AJB2, Q96I45
Tmem141
TMEM141 Fusion Protein Ag8341
150 μL
Primary
Human, Mouse, Rat