Learn More
TMEM35 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 24786-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16803285
  • 112.71€ / 20µL
Expédition estimée
03-03-2021
Spécification
TMEM35
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53FP2, Q9D328
TMEM35, transmembrane protein 35
Rabbit
IgG
20 μL
-20°C
Primary
59353, 67564
Western Blot
Unconjugated
TMEM35A
Liquid
TMEM35A
TMEM35 Fusion Protein Ag20589
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human