missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMIGD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27174-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
TMIGD1 (transmembrane and immunoglobulin domain containing 1) is a 262 amino acid single-pass membrane protein that contains two Ig-like C2-type (immunoglobulin-like) domains. It is a nonsecreted, glycosylated protein located in the cytoplasm and cell membrane. TMIGD1 might be a possible cell-differentiation marker in the intestine, whose expression is lost during cellular transformation and promote progression of tumorigenesis in the intestinal epithelium. TMIGD1 has also been postulated to play a role in cell adhesion.Spécification
TMIGD1 | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UXZ0, Q9D7L8 | |
TMIGD, TMIGD1, UNQ9372 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
388364, 66601 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TMIGD1 | |
Liquid | |
TMIGD1 | |
TMIGD1 Fusion Protein Ag25691 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |