missing translation for 'onlineSavingsMsg'
Learn More

TNFRSF17 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27724-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805795

  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is preferentially expressed in mature B lymphocytes, and may be important for B cell development and autoimmune response. This receptor has been shown to specifically bind to the tumor necrosis factor (ligand) superfamily, member 13b (TNFSF13B/TALL-1/BAFF), and to lead to NF-kappaB and MAPK8/JNK activation. This receptor also binds to various TRAF family members, and thus may transduce signals for cell survival and proliferation.
Spécification

Spécification

TNFRSF17
Polyclonal
Unconjugated
TNFRSF17
B cell maturation protein, BCM, BCMA, CD269, TNFRSF17
Rabbit
Antigen Affinity Chromatography
RUO
608
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02223
TNFRSF17
TNFRSF17 Fusion Protein Ag27049
20 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions