missing translation for 'onlineSavingsMsg'
Learn More

TRAF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26845-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805195

  • 112.71€ / 20µL

Expédition estimée
10-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the TNF receptor (TNFR) associated factor (TRAF) protein family. TRAF proteins associate with, and mediate the signal transduction from various receptors of the TNFR superfamily. This protein and TRAF2 form a heterodimeric complex, which is required for TNF-alpha-mediated activation of MAPK8/JNK and NF-kappaB. The protein complex formed by this protein and TRAF2 also interacts with inhibitor-of-apoptosis proteins (IAPs), and thus mediates the anti-apoptotic signals from TNF receptors. The expression of this protein can be induced by Epstein-Barr virus (EBV). EBV infection membrane protein 1 (LMP1) is found to interact with this and other TRAF proteins; this interaction is thought to link LMP1-mediated B lymphocyte transformation to the signal transduction from TNFR family receptors. Three transcript variants encoding two different isoforms have been found for this gene.
Spécification

Spécification

TRAF1
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13077
EBI6, TRAF1
Rabbit
IgG
20 μL
-20°C
Primary
7185
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Unconjugated
TRAF1
Liquid
TRAF1
TRAF1 Fusion Protein Ag25397
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents