Learn More
trans-4-Benzyl-N-Boc-L-proline, 95 %, Thermo Scientific™
Catalog Number p-7034450
Click to view available options
Quantité:
250 mg
1 g
153074-95-4
C17H23NO4
305.374
MFCD07784045
JPNHKKRLLDYCIZ-KGLIPLIRSA-N
2s,4r-4-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-2s,4r-4-benzyl-pyrrolidine-2-carboxylic acid, boc-trans-4-benzyl-l-proline, boc-r-gamma-benzyl-l-proline, 2s,4r-4-benzyl-1-2-methylpropan-2-yl oxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-benzyl pyrrolidine-2-carboxylic acid, 4beta-benzylpyrrolidine-1,2alpha-dicarboxylic acid 1-tert-butyl ester
2761964
(2S,4R)-4-benzyl-1-[(2-méthylpropan-2-yl)oxycarbonyl]pyrrolidine-2-acide carboxylique
CC(C)(C)OC(=O)N1CC(CC1C(=O)O)CC2=CC=CC=C2
153074-95-4
C17H23NO4
305.374
MFCD07784045
JPNHKKRLLDYCIZ-KGLIPLIRSA-N
2s,4r-4-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-2s,4r-4-benzyl-pyrrolidine-2-carboxylic acid, boc-trans-4-benzyl-l-proline, boc-r-gamma-benzyl-l-proline, 2s,4r-4-benzyl-1-2-methylpropan-2-yl oxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-benzyl pyrrolidine-2-carboxylic acid, 4beta-benzylpyrrolidine-1,2alpha-dicarboxylic acid 1-tert-butyl ester
2761964
(2S,4R)-4-benzyl-1-[(2-méthylpropan-2-yl)oxycarbonyl]pyrrolidine-2-acide carboxylique
CC(C)(C)OC(=O)N1CC(CC1C(=O)O)CC2=CC=CC=C2
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau trans-4-Benzyl-N-Boc-L-proline
Point de fusion 100°C to 102°C
Quantité 250 mg
Poids de la formule 305.37
Pourcentage de pureté 95%