missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRBV5-4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13524-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
TRBV5-4 | |
Polyclonal | |
Unconjugated | |
TRBV5-4 | |
TCRBV5S4, TCRBV5S6A3N2T, TRBV5 4, TRBV54, TRBV5-4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28611 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0A0C4DH59 | |
TRBV5-4 | |
TRBV5-4 Fusion Protein Ag4337 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |