Learn More
TRIM21 Mouse anti-Human, Unconjugated, Clone: 1E8B9, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67136-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The TRIM21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human, mouse, rat antigen. The TRIM21 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
TRIM21 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, Immunohistochemistry | |
1E8B9 | |
Unconjugated | |
20 μL | |
6737 | |
IgG1 |