missing translation for 'onlineSavingsMsg'
Learn More

TRIM22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13744-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802124

  • 112.71€ / 20µL

Date d’expédition estimée: 18-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

TRIM22 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. It localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the cytoplasm and its expression is induced by interferon. The protein down-regulates transcription from the HIV-1 LTR promoter region, suggesting that function of this protein may be to mediate interferon′s antiviral effects.
Spécification

Spécification

TRIM22
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IYM9
GPSTAF50, RING finger protein 94, RNF94, Staf 50, STAF50, TRIM22, tripartite motif containing 22
Rabbit
IgG
20 μL
-20°C
Primary
10346
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRIM22
Liquid
TRIM22
TRIM22 Fusion Protein Ag4683
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents