Learn More
TRIM59 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28575-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17281406
  • 349.00€ / 150µL
Expédition estimée: 04-10-2023
Description
TRIM59 belongs to the tripartite motif (TRIM) family. It functions as E3 ubiquitin ligases or an adaptor protein, and plays important roles in various types of human cancers.
Spécification
Spécification
TRIM59
Polyclonal
Unconjugated
150 μL
286827
Liquid
Western Blot, Immunofluorescence, ELISA
800 μg/mL
Rabbit
Primary
Human, Mouse, Rat