missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROAP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13634-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
TROAP | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12815 | |
TASTIN, TROAP, Trophinin assisting protein, Trophinin associated protein | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
10024 |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
TROAP | |
Liquid | |
TROAP | |
TROAP Fusion Protein Ag4562 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |