missing translation for 'onlineSavingsMsg'
Learn More

TRPC4AP Mouse anti-Human, Clone: 2C10B1, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66138-1-IG-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837745

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.
Spécification

Spécification

TRPC4AP
Monoclonal
0.96 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TEL6
TRPC4AP
TRPC4AP Fusion Protein Ag18772
20 μL
Primary
Human
Antibody
IgG3
Western Blot, Immunohistochemistry (Paraffin)
2C10B1
Unconjugated
TRPC4AP
C20orf188, Protein TAP1, Protein TRUSS, Trp4 associated protein, Trpc4 associated protein, TRPC4AP, TRRP4AP, TRUSS
Mouse
Protein A
RUO
26133
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions