missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21938-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.Spécification
TRPS1 | |
Polyclonal | |
Unconjugated | |
TRPS1 | |
GC79, LGCR, TRPS1, Zinc finger protein GC79 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299897, 7227, 83925 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q925H1, Q9UHF7 | |
TRPS1 | |
TRPS1 Fusion Protein Ag16583 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |